Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066642.6 | NP_499043.1 | 12 - 13 | 1808 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |