Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fozi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379857.1 | NP_001366672.1 | 3 - 4 | 419 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |