Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268073.3 | NP_001255002.1 | 5 - 6 | 513 | 88 | |
NM_001268074.4 | NP_001255003.1 | 5 - 6 | 575 | 88 | ||
GenBank mRNA | AB003924.1 | - | 4 - 5 | 440 | 88 | |
AB190513.1 | - | 4 - 5 | 440 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |