Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026054.4 | NP_001021225.1 | 13 - 14 | 4168 | 96 | |
NM_001268083.3 | NP_001255012.1 | 13 - 14 | 4227 | 96 | ||
NM_001379863.1 | NP_001366984.1 | 12 - 13 | 4065 | 96 | ||
NM_066714.4 | NP_499115.2 | 13 - 14 | 4170 | 96 | ||
GenBank mRNA | AF049142.1 | - | 13 - 14 | 4228 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |