Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306672.3 | NP_001293601.1 | 2 - 3 | 578 | 81 | |
NM_066775.6 | NP_499176.1 | 4 - 5 | 1121 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |