Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268104.2 | NP_001255033.1 | 1 - 2 | 108 | 129 | |
GenBank mRNA | AY324109.1 | - | 1 - 2 | 127 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |