Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mua-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027503.4 | NP_001022674.1 | 8 - 9 | 1408 | 77 | |
GenBank mRNA | AF139060.1 | - | 8 - 9 | 1408 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |