Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066873.6 | NP_499274.3 | 1 - 2 | 130 | 78 | |
GenBank mRNA | DQ104391.1 | - | 1 - 2 | 130 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |