Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027611.3 | NP_001022782.1 | 5 - 6 | 686 | 94 | |
NM_001379903.1 | NP_001368253.1 | 1 - 2 | 59 | 94 | ||
GenBank mRNA | AF151640.1 | - | 5 - 6 | 855 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |