Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027612.4 | NP_001022783.1 | 8 - 9 | 1336 | 95 | |
NM_001379901.2 | NP_001368252.1 | 8 - 9 | 1255 | 95 | ||
NM_001379902.1 | NP_001368254.1 | 4 - 5 | 626 | 95 | ||
NM_001379904.1 | NP_001367142.1 | 8 - 9 | 1336 | 95 | ||
NM_001382981.1 | 12 - 13 | 2151 | 95 | |||
GenBank mRNA | AF151641.1 | - | 8 - 9 | 1262 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |