Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027666.5 | NP_001022837.1 | 2 - 3 | 209 | 86 | |
NM_001382988.1 | 2 - 3 | 206 | 86 | |||
NM_001382989.1 | 2 - 3 | 185 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |