Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027482.4 | NP_001022653.1 | 4 - 5 | 630 | 57 | |
NM_001027483.3 | NP_001022654.1 | 4 - 5 | 621 | 57 | ||
GenBank mRNA | AF123573.1 | - | 4 - 5 | 615 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |