Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptr-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268156.3 | NP_001255085.1 | 6 - 7 | 830 | 63 | |
NM_001268157.3 | NP_001255086.1 | 6 - 7 | 802 | 63 | ||
NM_001268158.3 | NP_001255087.1 | 6 - 7 | 802 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |