Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hex-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066989.6 | NP_499390.3 | 2 - 3 | 797 | 138 |
Gene Symbol: Y39A1C.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382992.1 | 2 - 3 | 795 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |