Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: athp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066996.6 | NP_499397.2 | 10 - 11 | 2475 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |