Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: strd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027731.5 | NP_001022902.1 | 5 - 6 | 980 | 66 | |
NR_002379.1 | 5 - 6 | 954 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |