Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsks-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067046.4 | NP_499447.2 | 3 - 4 | 484 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |