Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129258.5 | NP_001122730.1 | 1 - 2 | 229 | 96 | |
NM_001379928.1 | NP_001367300.1 | 9 - 10 | 1078 | 96 | ||
NM_001379929.1 | NP_001367172.1 | 8 - 9 | 1408 | 96 | ||
NM_001382996.1 | 9 - 10 | 1155 | 96 | |||
NM_067047.6 | NP_499448.1 | 9 - 10 | 1226 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |