Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067105.5 | NP_499506.2 | 4 - 5 | 833 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |