Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067124.2 | NP_499525.1 | 3 - 4 | 445 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |