Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: top-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067157.6 | NP_499558.1 | 2 - 3 | 321 | 76 | |
GenBank mRNA | AF057032.1 | - | 2 - 3 | 321 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |