Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027777.4 | NP_001022948.1 | 1 - 2 | 232 | 82 | |
NM_001027778.5 | NP_001022949.1 | 1 - 2 | 230 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |