Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067198.5 | NP_499599.2 | 5 - 6 | 1133 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |