Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383010.1 | 3 - 4 | 335 | 60 | ||
NM_067201.6 | NP_499602.3 | 2 - 3 | 385 | 60 | ||
GenBank mRNA | AF160492.1 | - | 2 - 3 | 242 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |