Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383010.1 | 7 - 8 | 882 | 64 | ||
NM_067201.6 | NP_499602.3 | 6 - 7 | 932 | 64 | ||
GenBank mRNA | AF160492.1 | - | 6 - 7 | 789 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |