Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 13 - 14 | 2545 | 87 | |
NM_001268249.2 | NP_001255178.1 | 13 - 14 | 2552 | 87 | ||
NM_001312958.3 | NP_001299887.1 | 13 - 14 | 2561 | 87 | ||
NM_001312959.3 | NP_001299888.1 | 13 - 14 | 2554 | 87 | ||
NM_001312960.3 | NP_001299889.1 | 10 - 11 | 1975 | 87 | ||
NM_001312961.3 | NP_001299890.1 | 10 - 11 | 1975 | 87 | ||
NM_001312962.3 | NP_001299891.1 | 10 - 11 | 1966 | 87 | ||
NM_001312963.3 | NP_001299892.1 | 10 - 11 | 1966 | 87 | ||
NM_001312964.3 | NP_001299893.1 | 9 - 10 | 1405 | 87 | ||
NM_001312965.3 | NP_001299894.1 | 9 - 10 | 1405 | 87 | ||
NM_001312966.3 | NP_001299895.1 | 9 - 10 | 1396 | 87 | ||
NM_001312967.3 | NP_001299896.1 | 9 - 10 | 1396 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |