Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 18 - 19 | 3806 | 75 | |
NM_001268249.2 | NP_001255178.1 | 17 - 18 | 3723 | 75 | ||
NM_001312958.3 | NP_001299887.1 | 18 - 19 | 3822 | 75 | ||
NM_001312959.3 | NP_001299888.1 | 17 | 3725 | 75 | ||
NM_001312960.3 | NP_001299889.1 | 15 - 16 | 3236 | 75 | ||
NM_001312961.3 | NP_001299890.1 | 14 - 15 | 3146 | 75 | ||
NM_001312962.3 | NP_001299891.1 | 15 - 16 | 3227 | 75 | ||
NM_001312963.3 | NP_001299892.1 | 14 - 15 | 3137 | 75 | ||
NM_001312964.3 | NP_001299893.1 | 14 - 15 | 2666 | 75 | ||
NM_001312965.3 | NP_001299894.1 | 13 - 14 | 2576 | 75 | ||
NM_001312966.3 | NP_001299895.1 | 14 - 15 | 2657 | 75 | ||
NM_001312967.3 | NP_001299896.1 | 13 - 14 | 2567 | 75 | ||
NM_001312968.3 | NP_001299897.1 | 5 - 6 | 1238 | 75 | ||
NM_001312969.3 | NP_001299898.1 | 4 - 5 | 1148 | 75 | ||
NM_001312970.3 | NP_001299899.1 | 3 - 4 | 629 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |