Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 19 - 20 | 4001 | 80 | |
NM_001268249.2 | NP_001255178.1 | 18 - 19 | 3918 | 80 | ||
NM_001312958.3 | NP_001299887.1 | 19 - 20 | 4017 | 80 | ||
NM_001312959.3 | NP_001299888.1 | 18 - 19 | 3920 | 80 | ||
NM_001312960.3 | NP_001299889.1 | 16 - 17 | 3431 | 80 | ||
NM_001312961.3 | NP_001299890.1 | 15 - 16 | 3341 | 80 | ||
NM_001312962.3 | NP_001299891.1 | 16 - 17 | 3422 | 80 | ||
NM_001312963.3 | NP_001299892.1 | 15 - 16 | 3332 | 80 | ||
NM_001312964.3 | NP_001299893.1 | 15 - 16 | 2861 | 80 | ||
NM_001312965.3 | NP_001299894.1 | 14 - 15 | 2771 | 80 | ||
NM_001312966.3 | NP_001299895.1 | 15 - 16 | 2852 | 80 | ||
NM_001312967.3 | NP_001299896.1 | 14 - 15 | 2762 | 80 | ||
NM_001312968.3 | NP_001299897.1 | 6 - 7 | 1433 | 80 | ||
NM_001312969.3 | NP_001299898.1 | 5 - 6 | 1343 | 80 | ||
NM_001312970.3 | NP_001299899.1 | 4 - 5 | 824 | 80 | ||
NM_001379963.1 | NP_001368536.1 | 1 - 2 | 179 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |