Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268248.4 | NP_001255177.1 | 5 - 6 | 1251 | 104 | |
NM_001268249.2 | NP_001255178.1 | 5 - 6 | 1258 | 104 | ||
NM_001312958.3 | NP_001299887.1 | 5 - 6 | 1258 | 104 | ||
NM_001312959.3 | NP_001299888.1 | 5 - 6 | 1251 | 104 | ||
NM_001312960.3 | NP_001299889.1 | 2 - 3 | 672 | 104 | ||
NM_001312961.3 | NP_001299890.1 | 2 - 3 | 672 | 104 | ||
NM_001312962.3 | NP_001299891.1 | 2 - 3 | 672 | 104 | ||
NM_001312963.3 | NP_001299892.1 | 2 - 3 | 672 | 104 | ||
NM_001312964.3 | NP_001299893.1 | 1 - 2 | 102 | 104 | ||
NM_001312965.3 | NP_001299894.1 | 1 - 2 | 102 | 104 | ||
NM_001312966.3 | NP_001299895.1 | 1 - 2 | 102 | 104 | ||
NM_001312967.3 | NP_001299896.1 | 1 - 2 | 102 | 104 | ||
NM_001372667.1 | NP_001360800.1 | 5 - 6 | 1258 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |