Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067279.7 | NP_499680.1 | 2 - 3 | 850 | 80 | |
GenBank mRNA | U68185.1 | - | 2 - 3 | 850 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |