Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubq-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067294.5 | NP_499695.1 | 2 - 3 | 228 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |