Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lis-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067354.7 | NP_499755.1 | 7 - 8 | 955 | 75 | |
GenBank mRNA | AF164430.1 | - | 7 - 8 | 946 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |