Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067358.5 | NP_499759.2 | 4 - 5 | 918 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |