Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cua-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268273.3 | NP_001255202.1 | 16 - 17 | 3478 | 106 | |
NM_001268274.3 | NP_001255203.1 | 13 - 14 | 2979 | 106 | ||
GenBank mRNA | D83665.1 | - | 15 - 16 | 3414 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |