Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cua-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268273.3 | NP_001255202.1 | 5 - 6 | 741 | 166 | |
NM_001268274.3 | NP_001255203.1 | 2 - 3 | 242 | 166 | ||
GenBank mRNA | D83665.1 | - | 4 - 5 | 677 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |