Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cua-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268273.3 | NP_001255202.1 | 9 - 10 | 1550 | 95 | |
NM_001268274.3 | NP_001255203.1 | 6 - 7 | 1051 | 95 | ||
GenBank mRNA | D83665.1 | - | 8 - 9 | 1486 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |