Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kel-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067383.6 | NP_499784.2 | 10 - 11 | 1742 | 81 | |
NM_067384.6 | NP_499785.2 | 10 - 11 | 1727 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |