Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268276.2 | NP_001255205.1 | 5 - 6 | 738 | 139 | |
NM_001268277.1 | NP_001255206.1 | 5 - 6 | 754 | 139 | ||
NM_001268278.2 | NP_001255207.1 | 4 - 5 | 625 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |