Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027836.5 | NP_001023007.1 | 4 - 5 | 531 | 73 | |
NM_001027837.3 | NP_001023008.1 | 4 - 5 | 371 | 73 | ||
NM_001379988.1 | NP_001367020.1 | 3 - 4 | 342 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |