Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lit-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027634.3 | NP_001022805.1 | 6 - 7 | 763 | 98 | |
NM_001027635.5 | NP_001022806.1 | 4 - 5 | 428 | 98 | ||
NM_001027636.3 | NP_001022807.1 | 2 - 3 | 639 | 98 | ||
NM_001027637.5 | NP_001022808.1 | 7 - 8 | 795 | 98 | ||
NM_001379989.1 | NP_001367102.1 | 2 - 3 | 223 | 98 | ||
GenBank mRNA | AF143243.1 | - | 5 - 6 | 606 | 98 | |
AF143244.1 | - | 3 - 4 | 294 | 98 | ||
AF145376.1 | - | 6 - 7 | 763 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |