Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067446.7 | NP_499847.1 | 5 - 6 | 971 | 72 |
Gene Symbol: pdr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306674.3 | NP_001293603.1 | 4 - 5 | 758 | 72 | |
NM_001372678.3 | NP_001360308.1 | 4 - 5 | 871 | 72 | ||
NM_001383028.1 | 5 - 6 | 956 | 72 | |||
GenBank mRNA | AX254400.1 | - | 5 - 6 | 944 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |