Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372683.1 | NP_001359808.1 | 7 - 8 | 870 | 70 | |
NM_001372684.2 | NP_001359809.1 | 7 - 8 | 786 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |