Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col-99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129303.4 | NP_001122775.2 | 16 - 17 | 1893 | 72 | |
NM_001307632.3 | NP_001294561.1 | 14 - 15 | 1830 | 72 | ||
NM_001307633.3 | NP_001294562.1 | 14 - 15 | 1794 | 72 | ||
NM_001322765.3 | NP_001309608.1 | 13 - 14 | 1758 | 72 | ||
NM_001322767.3 | NP_001309606.1 | 14 - 15 | 1830 | 72 | ||
NM_001322769.3 | NP_001309607.1 | 14 - 15 | 1794 | 72 | ||
NM_001322771.3 | NP_001309605.1 | 15 - 16 | 1866 | 72 | ||
NM_001322773.3 | NP_001309602.1 | 13 - 14 | 1758 | 72 | ||
NM_067468.7 | NP_499869.3 | 15 - 16 | 1866 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |