Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col-99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129303.4 | NP_001122775.2 | 10 - 11 | 1158 | 66 | |
NM_001307632.3 | NP_001294561.1 | 8 - 9 | 1095 | 66 | ||
NM_001307633.3 | NP_001294562.1 | 9 - 10 | 1131 | 66 | ||
NM_001307634.4 | NP_001294563.1 | 9 - 10 | 1131 | 66 | ||
NM_001322764.3 | NP_001309612.1 | 8 - 9 | 1095 | 66 | ||
NM_001322765.3 | NP_001309608.1 | 8 - 9 | 1095 | 66 | ||
NM_001322766.3 | NP_001309610.1 | 8 - 9 | 1095 | 66 | ||
NM_001322767.3 | NP_001309606.1 | 8 - 9 | 1095 | 66 | ||
NM_001322768.3 | NP_001309611.1 | 9 - 10 | 1131 | 66 | ||
NM_001322769.3 | NP_001309607.1 | 9 - 10 | 1131 | 66 | ||
NM_001322770.3 | NP_001309609.1 | 9 - 10 | 1131 | 66 | ||
NM_001322771.3 | NP_001309605.1 | 9 - 10 | 1131 | 66 | ||
NM_001322772.3 | NP_001309604.1 | 8 - 9 | 1095 | 66 | ||
NM_001322773.3 | NP_001309602.1 | 8 - 9 | 1095 | 66 | ||
NM_001322774.3 | NP_001309603.1 | 8 - 9 | 1095 | 66 | ||
NM_001322775.2 | NP_001309601.1 | 9 - 10 | 1131 | 66 | ||
NM_067468.7 | NP_499869.3 | 9 - 10 | 1131 | 66 | ||
GenBank mRNA | KM875546.1 | - | 9 - 10 | 1131 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |