Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067482.4 | NP_499883.1 | 5 - 6 | 816 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |