Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqbp-1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067489.5 | NP_499890.2 | 2 - 3 | 349 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |