Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307480.3 | NP_001294409.1 | 14 - 15 | 2298 | 120 | |
NM_001307481.3 | NP_001294410.1 | 14 - 15 | 2298 | 120 | ||
NM_001307482.3 | NP_001294411.1 | 14 - 15 | 2298 | 120 | ||
NM_001307483.3 | NP_001294412.1 | 14 - 15 | 2298 | 120 | ||
NM_001307484.3 | NP_001294413.1 | 14 - 15 | 2298 | 120 | ||
NM_001307485.3 | NP_001294414.1 | 14 - 15 | 2298 | 120 | ||
NM_001307486.3 | NP_001294415.1 | 14 - 15 | 2298 | 120 | ||
NM_001307487.3 | NP_001294416.1 | 14 - 15 | 2298 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |