Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067520.9 | NP_499921.2 | 4 - 5 | 639 | 152 | |
GenBank mRNA | AY008127.1 | - | 4 - 5 | 600 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |