Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068730.1 | NP_501131.1 | 1 - 2 | 491 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |