Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307363.3 | NP_001294292.1 | 13 - 14 | 1390 | 118 | |
NM_001307364.3 | NP_001294293.1 | 12 - 13 | 1396 | 118 | ||
NM_001307365.3 | NP_001294294.1 | 11 - 12 | 1273 | 118 | ||
NM_068737.2 | NP_501138.1 | 13 - 14 | 1393 | 118 | ||
GenBank mRNA | U76402.1 | - | 13 - 14 | 1393 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |